Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Troponin T Type 3 (fast skeletal), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192534

Catalog No. NBP192534


Add to cart

Description

Description

Troponin T Type 3 (fast skeletal) Polyclonal antibody specifically detects Troponin T Type 3 (fast skeletal) in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Troponin T Type 3 (fast skeletal)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Beta-TnTF, fast skeletal muscle, TnTf, troponin T type 3 (skeletal, fast), troponin T3, skeletal, fast, troponin-T3, skeletal, fast
Rabbit
IgG
0.1 ml
Apoptosis, Cell Biology, Cytoskeleton Markers
Primary
7140
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:500
Affinity Purified
TNNT3
This antibody was developed against Recombinant Protein corresponding to amino acids:RKPLNIDHLGEDKLRDKAKELWETLHQLEIDKFEFGEKLKRQKYDITTLRSRIDQAQKHSKKAGTPAKGKVG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only