Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TROY Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA141061
Description
In Western blot, this antibody detects a band at ∽46 kDa (predicted molecular weight 50.8 kDa. For optimal results, primary antibody incubations should be performed at room temperature. Suggested positive control: antigen standard for TNFRSF19 (transient overexpression lysate), T98G.
TROY Polyclonal antibody specifically detects TROY in Human, Mouse samples. It is validated for Western Blot.Specifications
TROY | |
Polyclonal | |
Unconjugated | |
Tnfrsf19 | |
AL023044; AW123854; RGD1564996; TAJ; TAJ-alpha; TNF receptor superfamily member 19; Tnfrsf19; Toxicity and JNK inducer; TRADE; TROY; tumor necrosis factor receptor superfamily member 19; tumor necrosis factor receptor superfamily, member 19; UNQ1888/PRO4333 | |
Rabbit | |
Protein G | |
RUO | |
29820, 55504 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q9JLL3, Q9NS68 | |
Tnfrsf19 | |
Synthetic peptides corresponding to residues 29-44, 204-219, and 409-421 of human TROY. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction