Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRP Rabbit anti-Drosophila, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen OST00136G

Catalog No. PIOST00136G


Add to cart

Description

Description

Glycerol (1:1) may be added for additional stability.

Specifications

Specifications

TRP
Polyclonal
Unconjugated
trp
Lyophilized
TRP
A synthetic peptide from the 4th cytoplasmic loop of drosophila TRP.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Drosophila
Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
P19334
Transient receptor potential cation channel protein
Rabbit
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
43542
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit