Learn More
TRPM2 Rabbit anti-Mouse, Rat, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522884
Catalog No. PIPA522884
Add to cart
Description
Description
TRPM2 belongs to the ion transport protein family which includes Sodium, Potassium, Calcium ion channels. This family is characterized by proteins possessing 6 transmembrane helices in which the last two helices flank a loop which determines ion selectivity. TRPM2 is activated by oxidative stress and confers susceptibility to cell death. Further, TRPM2 is a calcium-permeable cation channel that is regulated by free intracellular ADP-ribose. Several alternatively spliced transcript variants of the TRPM2 gene have been described, but their full-length nature is not known.
Specifications
Specifications
TRPM2
Polyclonal
HRP
Trpm2
Liquid
TRPM2
Synthetic peptide made to an internal portion of the mouse protein (within residues 1200-1300).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse, Rat
Western Blot
1.5 mg/mL
PBS with no preservative
Q91YD4, E9PTA2
EREG1, KNP3, LTRPC2, NUDT9H, NUDT9L1, TRPC7, LTrpC-2, estrogen-responsive element-associated gene 1 protein, long transient receptor potential channel 2, transient receptor potential cation channel subfamily M member 2, transient receptor potential channel 7
Rabbit
IgG
100 μL
4° C, store in dark
Primary
28240, 294329