Learn More
TRPC7 Sheep anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Supplier:  Thermo Scientific OST00039W
Catalog No. PIOST00039W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water.
TRPC7 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC7 is probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. TRPC7 is also activated by diacylglycerol (DAG). Further, TRPC7 may also be activated by intracellular calcium store depletion.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPC7
Polyclonal
TRPC7
TRP7
Sheep
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q9HCX4, Q9WVC5
TRPC7
A synthetic peptide from the 1t cytoplasmic loop of human TRPC7 conjugated to an immunogenic carrier protein
RUO
26946, 282822, 57113
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions
Product Suggestions