Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPA1 Polyclonal Antibody, DyLight™ 550, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522835
Description
The target sequence has 69% sequence homology with rat. Suggested positive control: human brain membrane fraction.
TRPA1 Polyclonal antibody specifically detects TRPA1 in Guinea Pig, Human, Mouse, Rat, Zebrafish samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
TRPA1 | |
Polyclonal | |
DyLight 550 | |
Trpa1 | |
Anktm1; ankyrin-like with transmembrane domains 1; ankyrin-like with transmembrane domains protein 1; FEPS; ion channel; drTRPA1; nompC; transformation-sensitive protein p120; transient receptor potential A1; transient receptor potential ankyrin 1a; transient receptor potential cation channel subfamily A member 1; transient receptor potential cation channel subfamily A member 1a; transient receptor potential cation channel, subfamily A, member 1a; transient receptor potential cation channel subfamily A member 1-like; transient receptor potential cation channel, subfamily A, member 1; transient receptor potential cation channel, subfamily A, member 1 L homeolog; transient receptor potential cation channel, subfamily A, member 1a; TRPA1; TRPA-1; trpa1.L; trpa1a; TRPA1b; XELAEV_18003616mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100526649, 277328, 312896, 474351, 8989 | |
4° C, store in dark | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.68 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
O75762, Q6RI86, Q8BLA8 | |
Trpa1, trpa1a | |
A synthetic peptide made to a region within the N-terminus (residues 1-100) of the human TRPA1 protein. | |
100 μL | |
Primary | |
Guinea Pig, Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction