Learn More
TRPC2 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00074W
Catalog No. PIOSR00074W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Thought to form a receptor-activated calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Is not activated by intracellular calcium store depletion. Expressed exclusively in vomeronasal organ neurons (sensory microvilli). Belongs to the transient receptor family, STrpC subfamily. Contains 3 ANK repeats.
Specifications
Specifications
TRPC2
Polyclonal
whole serum with no preservative
Q9R283
Short transient receptor potential channel 2, rTRP2, Trp2, Trpc2 transient receptor potential cation channel, subfamily C, member 2, Trrp2
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunohistochemistry, Western Blot
Unconjugated
Trpc2
Lyophilized
TRPC2
A synthetic peptide from rat TRPC2 conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
64573