Learn More
TRPC2 Sheep anti-Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Supplier:  Thermo Scientific OST00021W
Catalog No. PIOST00021W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water.
Thought to form a receptor-activated calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Is not activated by intracellular calcium store depletion. Expressed exclusively in vomeronasal organ neurons (sensory microvilli). Belongs to the transient receptor family, STrpC subfamily. Contains 3 ANK repeats.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPC2
Polyclonal
Trpc2
TRPC2
A synthetic peptide from rat TRPC2, conjugated to an immunogenic carrier protein
RUO
64573
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot
Unconjugated
Q9R283
Sheep
100 μL
Primary
Rat
Antibody
IgG
Recommendations
Recommendations