Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPC2 Rabbit anti-Zebrafish, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen OST00078G

Catalog No. PIOST00078G


Add to cart

Description

Description

Reconstitute with 500 µL of distilled water.

Specifications

Specifications

TRPC2
Polyclonal
Unconjugated
trpc2b
TRPC2, Transient receptor potential channel C2
Rabbit
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
562120
Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
Lyophilized
trpc2b
A synthetic peptide from zebrafish TRPC2, conjugated to an immunogenic carrier protein
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Zebrafish
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit