Learn More
TRPC4AP Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OST00145W
Catalog No. PIOST00145W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability.
Specifications
Specifications
Trpc4ap
Polyclonal
whole serum with no preservative
Q8TEL6, Q9JLV2
Menthol receptorTrp4-associated protein, Short transient receptor potential channel 4-associated protein, TNF-receptor ubiquitous scaffolding/signaling protein, Protein TRUSS, TAP1 protein, Rabex-5/Rin2-interacting protein, TP4AP, Trpc4-associated protein, Trrp4ap
Rabbit
IgG
RUO
Antibody
Polyclonal
Chicken, Human, Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
TRPC4AP
Lyophilized
TRPC4AP
A synthetic peptide from chicken Trpc4ap
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26133, 362247, 428134, 56407