Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPC3/TRPC6/TRPC7 Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen OSR00067W

Catalog No. PIOSR00067W


Add to cart

Description

Description

The immunogenic sequence is homologous with the corresponding sequence derived from TRP6A, 6B and 6C proteins.

This is a G-protein-coupled receptor regulated Ca(2+) channel.
Specifications

Specifications

TRPC3/TRPC6/TRPC7
Polyclonal
whole serum with no preservative
Q9JMI9
Trpc3, Trpc6, Trpc7, Short transient receptor potential channel 3, Short transient receptor potential channel 6, Short transient receptor potential channel 7, Transient receptor protein 3, Transient receptor protein 6, Transient receptor protein 7
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunohistochemistry, Western Blot
Unconjugated
Trpc6
Lyophilized
TRPC3, TRPC6, TRPC7
A synthetic peptide from the 4th cytoplasmic domain of rat TRPC6, conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
282822, 60395, 89823
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit