Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPC3/TRPC6/TRPC7 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific OSR00068W
Description
Glycerol (1:1) may be added for additional stability.
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Activated by diacylglycerol (DAG) in a membrane-delimited fashion, independently of protein kinase C. Seems not to be activated by intracellular calcium store depletion.Specifications
TRPC3/TRPC6/TRPC7 | |
Polyclonal | |
TRPC7 | |
Trpc3, Trpc6, Trpc7, Short transient receptor potential channel 3, Short transient receptor potential channel 6, Short transient receptor potential channel 7, Transient receptor protein 3, Transient receptor protein 6, Transient receptor protein 7 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q13507, Q9HCX4, Q9Y210 | |
TRPC3, TRPC6, TRPC7 | |
A synthetic peptide from the 4th cytoplasmic loop of human TRPC6 conjugated to an immunogenic carrier protein | |
RUO | |
57113, 7222, 7225 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title