Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPC3/TRPC6/TRPC7 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen OSR00069W

Catalog No. PIOSR00069W


Add to cart

Description

Description

The immunogenic sequence shares 94% identity with TRPC3 and TRPC7 sequences.

Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Activated by diacylglycerol (DAG) in a membrane-delimited fashion, independently of protein kinase C. Seems not to be activated by intracellular calcium store depletion.
Specifications

Specifications

TRPC3/TRPC6/TRPC7
Polyclonal
whole serum with no preservative
Q61143, Q9QZC1, Q9WVC5
Short transient receptor potential channel 6, TrpC6, Calcium entry channel, Trp6, transient receptor potential cation channel, subfamily C, member 6,
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Trpc3
Lyophilized
TRPC3, TRPC6, TRPC7
A synthetic peptide from the 4th cytoplasmic loop of mouse TRPC6 conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
22065, 22068, 26946
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit