Learn More
TRPM1 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00048W
Catalog No. PIOSR00048W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material. OSR00048W can be used with blocking peptide GST00008P.
The protein encoded by this gene is similar to the transient receptor potential (TRP) calcium channel family members. The expression of this protein is inversely correlated with melanoma aggressiveness, suggesting that it suppresses melanoma metastasis.
Specifications
Specifications
TRPM1
Polyclonal
whole serum with no preservative
Q2WEA5
Transient receptor potential cation channel,subfamily M, member 1 variant delta1066-1071, Trpm1, LTRPC1, MLSN1, melastatin
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunohistochemistry, Western Blot
Unconjugated
Trpm1
Lyophilized
TRPM1
A synthetic peptide from rat TRPM1 conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
361586