Learn More
TRPM8 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00077W
Catalog No. PIOSR00077W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
TRPM8 is a receptor-activated non-selective cation channel involved in detection of sensations such as coolness, by being activated by cold temperature below 25 degrees Celsius. TRPM8 is activated by icilin, eucalyptol, menthol, cold and modulation of intracellular pH and involved in menthol sensation. TRPM8 is permeable for monovalent cations sodium, potassium, and cesium and divalent cation calcium. TRPM8 has an expression pattern within normal human tissues that is mostly restricted to prostate epithelial cells. TRPM8 mRNA expression is at moderate levels in normal prostate tissue and appears to be elevated in prostate cancer. Notably, TRPM8 mRNA was also expressed in a number of nonprostatic primary tumors of breast, colon, lung, and skin origin, whereas transcripts encoding TRPM8 were hardly detected or not detected in the corresponding normal human tissues.
Specifications
Specifications
TRPM8
Polyclonal
whole serum with no preservative
Q8R455
Transient receptor potential cation channel subfamily M member 8, Cold menthol receptor 1, Trpm8, Trpm8 transient receptor potential cation channel, s
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Trpm8
Lyophilized
TRPM8
A synthetic peptide from the 4th cytoplasmic loop of rat TRPM8, conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
171384