Learn More
TRPV2 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00060W
Catalog No. PIOSR00060W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material. OSR00060W can be used with blocking peptide GST00010P.
Calcium-permeable, non-selective cation channel with an outward rectification. Seems to be regulated, at least in part, by growth factors, like IGF1, PDGF and morphogenetic neuropeptide/head activator. May transduce physical stimuli in mast cells. Activated by temperatures higher than 52 degrees Celsius; is not activated by vanilloids and acidic pH. Interacts with a cAMP-dependent protein kinase type II regulatory subunit (PRKAR2A or PRKAR2B) and ACBD3. Interacts with RGA. Ubiquitously expressed. Expressed in dorsal root ganglia, trigeminal ganglia, spinal chord (Lissauer ft.s tract, dorsal horn and dorsal columns) (at protein level).
Specifications
Specifications
TRPV2
Polyclonal
whole serum with no preservative
Q9WUD2
Transient receptor potential cation channel subfamily V member 2, TrpV2, osm-9-like TRP channel 2, OTRPC2, Vanilloid receptor-like protein 1, VRL-1, S
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunohistochemistry, Western Blot
Unconjugated
Trpv2
Lyophilized
TRPV2
A synthetic peptide from the 3d extracellular loop of rat TRPV2
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
29465