Learn More
TRPV3 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00056W
Catalog No. PIOSR00056W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material. OSR00056W can be used with blocking peptide GST00011P.
Putative receptor-activated non-selective calcium permeant cation channel. It is activated by innocuous (warm) temperatures and shows an increased response at noxious temperatures greater than 39 degrees Celsius. Activation exhibits an outward rectification. May associate with TRPV1 and may modulate its activity. Interacts with TRPV1. Expressed in keratinocytes and hair follicles Belongs to the transient receptor family, TrpV subfamily. Contains 3 ANK repeats.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPV3
Polyclonal
Trpv3
TRPV3
A synthetic peptide the 1t cytoplasmic domain of rat TRPV3 conjugated to an immunogenic carrier protein
RUO
497948
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot
Unconjugated
Heat sensitive channel TRPV3
Rabbit
100 μL
Primary
Rat
Antibody
IgG
Recommendations
Recommendations