Learn More
TRPV3 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00057W
Catalog No. PIOSR00057W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Putative receptor-activated non-selective calcium permeant cation channel. It is activated by innocuous (warm) temperatures and shows an increased response at noxious temperatures greater than 39 degrees Celsius. Activation exhibits an outward rectification. May associate with TRPV1 and may modulate its activity. Interacts with TRPV1. Expressed in keratinocytes and hair follicles Belongs to the transient receptor family, TrpV subfamily. Contains 3 ANK repeats.
Specifications
Specifications
TRPV3
Polyclonal
whole serum with no preservative
Q8K424
Trpv3 transient receptor potential cation channel, subfamily V, member 3, Nh, VRL3, AI644701, MGC124324, MGC124325, 1110036I10Rik
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Trpv3
Lyophilized
TRPV3
A synthetic peptide from the 1t cytoplasmic domain of mouse TRPV3 conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
246788