Learn More
TRPV3 Sheep anti-Mouse, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen OST00007W
Catalog No. PIOST00007W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
TRPV3 belongs to a family of nonselective cation channels that function in a variety of processes, including temperature sensation and vasoregulation. The thermosensitive members of this family are expressed in subsets of sensory neurons that terminate in the skin, and are activated at distinct physiological temperatures. This channel is activated at temperatures between 22 and 40 degrees C. This gene lies in close proximity to another family member (TRPV1) gene on chromosome 17, and the two encoded proteins are thought to associate with each other to form heteromeric channels.
Specifications
Specifications
TRPV3
Polyclonal
whole serum with no preservative
Q8K424
VRL3
Sheep
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Trpv3
Lyophilized
TRPV3
A synthetic peptide from mouse TRPV3 conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
246788