Learn More
TRPV5 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00129W
Catalog No. PIOSR00129W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
This gene is a member of the transient receptor family and the TrpV subfamily. The calcium-selective channel encoded by this gene has 6 transmembrane-spanning domains, multiple potential phosphorylation sites, an N-linked glycosylation site, and 5 ANK repeats. This protein forms homotetramers or heterotetramers and is activated by a low internal calcium level. Constitutively active calcium selective cation channel thought to be involved in Ca(2+) reabsorption in kidney and intestine. The channel is activated by low internal calcium level and the current exhibits an inward rectification. A Ca(2+)-dependent feedback regulation includes fast channel inactivation and slow current decay. Heteromeric assembly with TRPV6 seems to modify channel properties. TRPV5-TRPV6 heteromultimeric concatemers exhibit voltage-dependent gating. Colocalized with S100A10 and ANAX2 along the apical domain of kidney distal tubular cells. Expressed at high levels in kidney, small intestine and pancreas, and at lower levels in testis, prostate, placenta, brain, colon and rectum.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPV5
Polyclonal
TRPV5
TrpV5, osm-9-like TRP channel 3, OTRPC3, Epithelial calcium channel 1, ECaC, ECaC1, Calcium transport protein 2, CaT2, CAT2, ECAC1, OTRPC3, transient
Rabbit
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q9NQA5
TRPV5
A synthetic peptide from the 4th cytoplasmic loop of human TRPV5 conjugated to an immunogenic carrier protein
RUO
56302
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions
Product Suggestions