Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-TrpV6 Clone: 4A5.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABN839
Description
Specifically detects TrpV6 Clone: 4A5.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting
Transient receptor potential cation channel subfamily V member 6 (UniProt Q9H1D0; also known as Alu-binding protein with zinc finger domain, Calcium transport protein 1, Calcium transporter-like protein, CaT-L, CaT-like, CaT1, ECaC2, Epithelial apical membrane calcium transporter/channel CaT1, Epithelial calcium channel 2, TrpV6) is encoded by the TRPV6 (also known as ABP/ZF, CAT1, CATL, ECAC2, ZFAB) gene (Gene ID 55503) in human. TRPV6 belongs to the transient receptor potential (TRP) family of membrane proteins. The human TRPV6 is an 765-amino acid, six-transmembrane protein with both N- and C-termini exposed intracellularly. TRPV6 contains a pore-forming loop domain located between the fifth and sixth transmembrane region. Among all the TRPV family members, TRPV5 and TRPV6 are the two highly Ca2+-selective channels that play pivotal roles in the Ca2+-reabsorption in the kidney and intestine, respectively. TRPV6 is found in the apical brush-border membrane of the intestinal enterocytes where it regulates calcium entry into the cells. TRPV6 expression is also reported in human placenta, pancreas, and prostate gland. Vitamin D treatment of human colon cancer Caco-2 cells and duodenal explants results in upregulated TRPV6 mRNA levels.
Specifications
TrpV6 | |
Monoclonal | |
Unconjugated | |
Q9H1D0 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
Expected to react with both spliced isoforms of human TrpV6 reported by UniProt (Q9H1D0). | |
-20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles | |
IgG2a κ |
Immunohistochemistry, Western Blot | |
4A5.1 | |
In culture supernatant without preservatives. | |
TRPV6, ABP/ZF, CAT1, CATL, ECAC2, ZFAB | |
GST-tagged recombinant human TrpV6 C-terminal fragment. | |
100 μL | |
Neuroscience | |
NP_061116 | |
Human, Mouse, Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction