Learn More
TRPV6 Sheep anti-Human, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen OST00047W
Catalog No. PIOST00047W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Calcium-permeable channels, such as TRPV6, participate in neurotransmission, muscle contraction, and exocytosis by providing calcium as an intracellular second messenger. Depending on the tissue, transcellular calcium transport may be regulated by vitamin D, parathyroid hormone (PTH), or calcitonin (CALCA). Calcium selective cation channel probably involved in Ca(2+) uptake in various tissues, including Ca(2+) reabsorption in intestine. The channel is activated by low internal calcium level, probably including intracellular calcium store depletion, and the current exhibits an inward rectification. Inactivation includes both, a rapid Ca(2+)-dependent and a slower Ca(2+)-calmodulin-dependent mechanism, the latter may be regulated by phosphorylation. In vitro, is slowly inhibited by Mg(2+) in a voltage-independent manner. Heteromeric assembly with TRPV5 seems to modify channel properties. TRPV5-TRPV6 heteromultimeric concatemers exhibit voltage-dependent gating. Interacts with TRPV5. Interacts with S100A10 and probably with the ANAX2-S100A10 heterotetramer. The interaction with S100A10 is required for the trafficking to the plasma membrane. Interacts with BSPRY. Interacts with calmodulin.
Specifications
Specifications
TRPV6
Polyclonal
whole serum with no preservative
Q9H1D0
ABP/ZF, CAT1, CATL, ECAC2, HSA277909, LP6728, ZFAB
Sheep
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
TRPV6
Lyophilized
TRPV6
A synthetic peptide from human TRPV6 conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
55503