Learn More
TRPV6 Rabbit anti-Zebrafish, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OST00067W
Catalog No. PIOST00067W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Calcium-permeable channels, such as TRPV6, participate in neurotransmission, muscle contraction, and exocytosis by providing calcium as an intracellular second messenger. Depending on the tissue, transcellular calcium transport may be regulated by vitamin D, parathyroid hormone (PTH; MIM 168450), or calcitonin (CALCA; MIM 114130).
Specifications
Specifications
TRPV6
Polyclonal
whole serum with no preservative
Lyophilized
TRPV6
A synthetic peptide from zebrafish TRPV6, conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100000322
Immunohistochemistry, Western Blot
Unconjugated
trpv6
trpv6, epithelial calcium channel, transient receptor potential cation channel, subfamily V, member 6
Rabbit
IgG
RUO
Antibody
Polyclonal
Zebrafish