Learn More
Trypsin Inhibitor Rabbit anti-Plant, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA127229
Catalog No. PA127229
Add to cart
Description
Description
PA1-27229 detects Trypsin Inhibitor from soy bean samples. PA1-27229 has been successfully used in immunohistochemistry, ELISA, and Western blot procedures. The PA1-27229 immunogen is Trypsin Inhibitor from soy bean.
Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screen for and diagnose Cystic Fibrosis.
Specifications
Specifications
Trypsin Inhibitor
Polyclonal
HRP
Liquid
Trypsin Inhibitor from soy bean.
IgG fraction
RUO
Antibody
Polyclonal
ELISA, Immunohistochemistry, Immunomicroscopy, Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2
Rabbit
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Plant