Learn More
Trypsin Mouse anti-Human, Clone: TRYP11-224.3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110616
Catalog No. 01-671-569
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10616 detects Trypsin from human samples. MA1-10616 has been successfully used in ELISA applications. The MA1-10616 immunogen is purified human pancreatic Trypsin.
This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is secreted by the pancreas and cleaved to its active form in the small intestine. It is active on peptide linkages involving the carboxyl group of lysine or arginine. Mutations in this gene are associated with hereditary pancreatitis. This gene and several other trypsinogen genes are localized to the T cell receptor beta locus on chromosome 7.
Specifications
Specifications
Trypsin
Monoclonal
0.87 mg/mL
PBS with no preservative; pH 7.4
P07477
MGC120175, MGC149362, TRP1, TRY1, TRY4, TRYP1
Mouse
IgG2b
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5644
ELISA
TRYP11-224.3
Unconjugated
PRSS1
Liquid
PRSS1
Purified human pancreatic Trypsin.
Protein G
RUO
Antibody
Monoclonal
Human