Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPSB2 Monoclonal Antibody (349406), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524090
Description
In direct ELISAs and Western blots, 50-100% cross-reactivity with recombinant human (rh) TPSAB1 and recombinant mouse (rm) Mcpt6 is observed, 20-100% cross-reactivity with rmMcpt7 is observed, and no cross-reactivity with rhTPSG1, rmTPS5, rmMcpt1, or rmMcpt11 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
TPSB2 Monoclonal antibody specifically detects TPSB2 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
TPSB2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P20231 | |
Tpsb2 | |
Mouse myeloma cell line NS0-derived recombinant human Tryptase beta-2/TPSB2 Ile31-Pro275 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
349406 | |
Unconjugated | |
Tpsb2 | |
AV011504; EC 3.4.21; EC 3.4.21.59; mast cell protease 6; mast cell tryptase beta II; mast cell tryptase beta III; Mcp6; Mcp-6; Mcpt6; MMCP-6; rMCP-6; testicular tissue protein Li 163; TPS2; TPS2tryptase beta-2; Tpsb2; tryptase beta 2; tryptase beta 2 (gene/pseudogene); tryptase beta II; tryptase beta III; tryptase beta-2; Tryptase II; tryptase III; Tryptase-2; tryptaseB; tryptaseC; Tryptase-C | |
Mouse | |
Protein A/G | |
RUO | |
64499 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction