Learn More
TSCOT Mouse anti-Human, Clone: 2F7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522417
Catalog No. PIMA522417
Add to cart
Description
Description
Peptide Sequence: LKVPESVAKP SQELPAVDTV SGTVGTYRTL DPDQLDQQYA VGHPPSPGKA KPHKTT
May act as a transporter.
Specifications
Specifications
TSCOT
Monoclonal
1.0 mg/mL
PBS with no preservative; pH 7.4
Q9BY10
Ly110, TSCOT, thymic stromal co-transporter, thymic stromal cotransporter homolog
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
57864
ELISA, Western Blot
2F7
Unconjugated
SLC46A2
Liquid
SLC46A2
SLC46A2 (NP_149040, 227 a.a. ∽ 282 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human