Learn More
TSKS Mouse anti-Human, Clone: 2E9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522438
Catalog No. PIMA522438
Add to cart
Description
Description
Peptide Sequence: ALTSLVDEVK QRGLTPACPS CQRLHKKILE LERQALAKHV RAEALSSTLR LAQDEALRAK NLLLTDKMKP EEKMATLDHL HLKMCSLHDH
This gene may play a role in testicular physiology, spermatogenesis or spermiogenesis. Expression of the encoded protein is highest in the testis and down-regulated in testicular cancer. The gene is localized to the region 19q13.3 among the related RAS viral oncogene homolog (RRAS) and interferon regulatory factor 3 (IRF3) genes, which are both involved in tumorigenesis pathways and progression.
Specifications
Specifications
TSKS
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9UJT2
STK22S1, TSKS1, TSSKS, STK22 substrate 1, testis specific serine/threonine kinase substrate, testis-specific kinase substrate
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
60385
ELISA, Western Blot
2E9
Unconjugated
TSKS
Liquid
TSKS
TSKS (NP_068379, 471 a.a. ∽ 560 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human