Learn More
Tspan-27 (CD82) Mouse anti-Human, Alexa Fluor 647, Clone: 423524, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564341
Catalog No. BDB564341
Add to cart
Description
Description
The 423524 monoclonal antibody specifically binds to Tetraspanin-27 (Tspan-27) which is also known as CD82, R2, C33, KAI1, 4F9, IA4, and ST6. The CD82 glycoprotein is a member of tetraspanin superfamily that has four putative transmembrane domains. CD82 is widely expressed on a variety of cell types including granulocytes, monocytes, fibroblasts, epithelial cells, endothelial cells, CD34+ progenitor cells, and at varying levels on T cell and B cell subsets. CD82 associates with other surface molecules including integrins, MHC molecules, CD37, CD53, and CD81 and forms signaling receptor complexes. These can regulate cellular responses including the activation of monocytes and costimulation of T cells. CD82 appears to play a role in the suppression of tumor cell metastasis.
Specifications
Specifications
Tspan-27 (CD82)
423524
KAI1; 4F9; C33; IA4; R2; SAR2; ST6; GR15; Tspan-27; Tetraspanin-27
Affinity Purified
Human CD82 Transfected Cell Line
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human