Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526329

Catalog No. PIPA526329


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

TSPAN2 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
Specifications

Specifications

TSPAN2
Polyclonal
PBS with 0.09% sodium azide
O60636
NET3, TSN2, TSPAN-2, new EST tetraspan 3, tetraspan 2, tetraspan NET-3, tetraspan TM4SF, tetraspanin 2 isoform, tetraspanin-2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10100
Flow Cytometry, Western Blot
Unconjugated
TSPAN2
Liquid
TSPAN2
KLH conjugated synthetic peptide between 109-137 amino acids from the central region of human TSPAN2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit