Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525472
Description
This antibody is predicted to react with bovine based on sequence homology.
TSPAN3 Polyclonal antibody specifically detects TSPAN3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TSPAN3 | |
Polyclonal | |
Unconjugated | |
TSPAN3 | |
1700055K04Rik; H41D4 protein; OAP-1; OSP-associated protein 1; tetraspan 3; tetraspan TM4SF; tetraspanin 3; tetraspanin TM4-A; tetraspanin TM4-A homolog; tetraspanin-3; TM4-A; Tm4sf8; Transmembrane 4 superfamily member 8; TSPAN3; TSPAN-3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10099, 56434 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O60637, Q9QY33 | |
TSPAN3 | |
KLH conjugated synthetic peptide between 90-117 amino acids from the central region of human TSPAN3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction