Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TSTA3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133070

Catalog No. NBP133070


Add to cart

Description

Description

TSTA3 Polyclonal antibody specifically detects TSTA3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TSTA3
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
TSTA3
Recombinant protein encompassing a sequence within the C-terminus region of human TSTA3. The exact sequence is proprietary.
36 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
EC 1.1.1.271, FX, GDP-4-keto-6-deoxy-D-mannose epimerase-reductase, GDP-4-keto-6-deoxy-D-mannose-3,5-epimerase-4-reductase, GDP-L-fucose synthase, P35B, Protein FX, Red cell NADP(H)-binding protein, SDR4E13-5 epimerase/4-reductase, short chain dehydrogenase/reductase family 4E, member 1, Short-chain dehydrogenase/reductase family 4E member 1, tissue specific transplantation antigen 3, tissue specific transplantation antigen P35B, Tissue-specific transplantation antigen-3
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
7264
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only