Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC26 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525245
Description
This antibody is predicted to react with mouse, non-human primate and rat based on sequence homology.
Specifications
TTC26 | |
Polyclonal | |
TTC26 | |
DYF13, dyf-13, TPR repeat protein 26, tetratricopeptide repeat protein 26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79989 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
A0AVF1 | |
TTC26 | |
KLH conjugated synthetic peptide between 467-496 amino acids from the C-terminal region of human TTC26 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction