Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC30B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524518

Catalog No. PIPA524518

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and Xenopus and zebrafish based on sequence homology.

Required for polyglutamylation of axonemal tubulin (By similarity). Plays a role in anterograde intraflagellar transport (IFT), the process by which cilia precursors are transported from the base of the cilium to the site of their incorporation at the tip.
Specifications

Specifications

TTC30B
Polyclonal
PBS with 0.09% sodium azide
Q8N4P2, Q9CY00
IFT70, fleer, TPR repeat protein 30B, tetratricopeptide repeat protein 30B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
150737, 72421
Western Blot
Unconjugated
TTC30B
Liquid
TTC30B
KLH conjugated synthetic peptide between 554-582 amino acids from the C-terminal region of human TTC30B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit