Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tetratricopeptide repeat domain 4 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TTC4 protein.
Supplier: Abnova Corporation H00007268B01
Description
The 34-amino acid tetratricopeptide repeat (TPR) motif is found in a variety of proteins and may mediate protein-protein or protein-membrane interactions.[supplied by OMIM
Sequence: MEQPGQDPTSDDVMDSFLEKFQSQPYRGGFHEDQWEKEFEKVPLFMTRAPSEIDPRENPDLACLQSIIFDEERSPEEQAKTYKDEGNDYFKEKDYKKAVISYTEGLKKKCADPDLNAVLYTNRAAAQYYLGNFRSALNDVTAARKLKPCHLKAIIRGALCHLELKHFAEAVNWCDEGLQIDAKEKKLLEMRAKADKLKRIEQRDVRKANLKEKKERNQNEALLQAIKARNIRLSEAACEDEDSASEGLGELFLDGLSTENPHGARLSLDGQGRLSWPVLFLYPEYAQSDFISAFHEDSRFIDHLMVMFGETPSWDLEQKYCPDNLEVYFEDEDRAELYRVPAKSTLLQVLQHQRYFVKALTPAFLVCVGSSPFCKNFLRGRKVYQIRSpecifications
tetratricopeptide repeat domain 4 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TTC4 protein. | |
TTC4 | |
DKFZp781B0622/FLJ41930/MGC5097 | |
Mouse | |
50 μL | |
Yes | |
7268 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC001276 | |
TTC4 | |
TTC4 (AAH01276, 1 a.a. ∼ 387 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction