Learn More
tetratricopeptide repeat domain 8, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TTC8 protein.
Manufacturer: Abnova Corporation H00123016B01
Description
This gene encodes a protein that has been directly linked to Bardet-Biedl syndrome. The primary features of this syndrome include retinal dystrophy, obesity, polydactyly, renal abnormalities and learning disabilities. Experimentation in nonhuman eukaryotes suggests that this gene is expressed in ciliated cells and that it is involved in the formation of cilia. Alternate transcriptional splice variants have been characterized. [provided by RefSeq
Sequence: MSSEMEPLLLAWSYFRRRKFQLCADLCTQMLEKSPYDQAAWILKARALTEMVYIDEIDVDQEGIAEMMLDENAIAQVPRPGTSLKLPGTNQTGGPSQAVRPITQAGRPITGFLRPSTQSGRPGTMEQAIRTPRTAYTARPITSSSGRFVRLGTASMLTSPDGPFINLSRLNLTKYSQKPKLAKALFEYIFHHENDVKTALDLAALSTEHSQYKDWWWKVQIGKCYYRLGMYREAEKQFKSALKQQEMVDTFLYLAKVYVSLDQPVTALNLFKQGLDKFPGEVTLLCGIARIYEEMNNMSSAAEYYKEVLKQDNTHVEAIACIGSNHFYSDQPEIALRFYRRLLQMGIYNGQLFNNLGLCCFYAQQYDMTLTSFERALSLAENEEEAADVWYNLGHVAVGIGDTNLAHQCFRLALVNNNNHAEAYNNLAVLEMRKGHVEQARALLQTASSLAPHMYEPHFNFATISDKIGDLQRSYVAAQKSEAAFPDHVDTQHLIKQLRQHFAMLSpecifications
tetratricopeptide repeat domain 8 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TTC8 protein. | |
TTC8 | |
BBS8 | |
Mouse | |
50 μL | |
Yes | |
123016 | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_198309.2 | |
TTC8 | |
TTC8 (NP_938051.1, 1 a.a. ∼ 505 a.a) full-length human protein. | |
RUO | |
Primary | |
Human |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.