Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTPAL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525088

Catalog No. PIPA525088


Add to cart

Description

Description

TTPAL may act as a protein that binds a hydrophobic ligand.
Specifications

Specifications

TTPAL
Polyclonal
PBS with 0.09% sodium azide
Q9BTX7
C20orf121, alpha-tocopherol transfer protein-like
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79183
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
TTPAL
Liquid
TTPAL
KLH conjugated synthetic peptide between 204-233 amino acids from the C-terminal region of human TTPAL
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit