Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TULP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521534

Catalog No. PIPA521534


Add to cart

Description

Description

PA5-21534 targets Tubby-related protein 1 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-21534 immunogen is a recombit fragment corresponding to a region within amino acids 281 and 542 of TULP1 (Uniprot IDNo.O00294). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

TULP1 is a member of the tubby-like gene family (TULPs). Members of this family have been identified in plants, vertebrates, and invertebrates. TULP proteins share a conserved C-terminal region of approximately 200 amino acid residues. The protein encoded by this gene is thought to play a role in the physiology of photoreceptors. Mutations in this gene are associated with recessive juvenile retinitis pigmentosa and Leber congenital amaurosis-15.
Specifications

Specifications

TULP1
Polyclonal
Unconjugated
TULP1
Liquid
TULP1
Recombit fragment corresponding to a region within amino acids 281 and 542 of TULP1 (Uniprot IDNo.O00294)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.45 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O00294
RP3-340B19.1, LCA15, RP14, TUBL1, tubby-related protein 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7287
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit