Learn More
α-Tubulin Mouse anti-Avian, Bovine, Guinea Pig, Human, Mouse, Pig, Rat, AlexaFluor™555, Clone: DM1A, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05829X555
Catalog No. 05-829-X555MI
Add to cart
Description
Description
Anti-α-Tubulin Antibody, Alexa Fluor 555 conjugate detects level of α-Tubulin and has been published and validated for use in immunofluoresence.
Specifications
Specifications
α-Tubulin
DM1A
Protein G purified immunoglobulin conjugated to Alexa Fluor™ 555. Liquid in PBS containing 1% BSA, 0.05% Tween™-20 and 0.05% sodium azide.
Purified
Murine
IgG1
100μg
Primary
Avian, Bovine, Guinea Pig, Human, Murine, Porcine, Rat
Immunofluorescence
Alexa Fluor 555
NM_006009
TUBA1A, hum-a-tub2, FLJ25113, B-ALPHA-1, TUBA3, hum-a-tub1
The epitope recognized by this antibody has been mapped to amino acids 426-450 (AALEKDYEEVGVDSV-EGEGEEEGEE).
Protein G purfied
4°C for up to 12 months
Monoclonal