Learn More
Β-Tubulin Mouse anti-Human, Rat, Mouse, Bovine, Clone: AA2, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05661
Catalog No. 05-661-MI
Add to cart
Description
Description
This Anti-β-Tubulin Antibody, clone AA2 is validated for use in western blotting for the detection of β-Tubulin.
Specifications
Specifications
β-Tubulin
AA2
Purified mouse monoclonal IgG1 in buffer containing PBS with 0.05% sodium azide and 30% glycerol.
Purified
Murine
IgG1
200μg
Primary
Bovine, Human, Murine, Rat
Western Blot
Unconjugated
NM_178014
M40, MGC117247, MGC16435, OK/SW-cl.56, OTTHUMP00000029069, TUBB1, TUBB5, Tubb5
Purified bovine brain tubulin. The epitope recognized by this antibody has been mapped to amino acids 412-430.
Protein G purfied
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal