Learn More
Tubulin ΒIII Mouse anti-Bovine, Human, Mouse, Rat, Clone: 2G10, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05559
Catalog No. 05-559-MI
Add to cart
Description
Description
Anti-Tubulin βIII Antibody, clone 2G10, neuronal is an antibody against Tubulin βIII for use in western blotting and immunocytochemistry.
Specifications
Specifications
Tubulin βIII
2G10
PBS, pH 7.4, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
IgG2a
200μg
Primary
Bovine, Human, Murine, Rat
Immunocytochemistry, Western Blot
Unconjugated
NM_030773
TUBB3, TUBB4, beta-4, MC1R
Peptide corresponding to residues 436-450 of neuronal specific βIII tubulin; epitope mapped to EAQGPK
Protein G chromatography
−20°C for 24 months.
Monoclonal