Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha/beta Tubulin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA186602
Description
Rabbit anti Porcine tubulin antibody recognizes tubulin, a protein which is the major constituent of microtubules.
alpha/beta Tubulin Polyclonal antibody specifically detects alpha/beta Tubulin in Human, Mouse, Porcine, Rat samples. It is validated for ELISA, Western Blot.Specifications
alpha/beta Tubulin | |
Polyclonal | |
Unconjugated | |
TUBB | |
2810484G07Rik; Alpha-tubulin 1; Alpha-tubulin 3; Alpha-tubulin isotype M-alpha-1; B-ALPHA-1; beta 5-tubulin; beta Ib tubulin; CDCBM6; CSCSC1; Detyrosinated tubulin alpha-1A chain; FLJ30169; H2 ALPHA; hum-a-tub1; hum-a-tub2; LIS3; M(beta)1; M40; OK/SW-cl.56; Talpha1 alpha-tubulin; Tuba1; Tuba-1; TUBA1A; TUBA3; TUBB; TUBB1; TUBB2A; TUBB5; Tubulin alpha; tubulin alpha 1a; tubulin alpha 3; tubulin alpha-1 chain; Tubulin alpha-1A chain; tubulin alpha-3 chain; Tubulin B-alpha-1; tubulin beta; tubulin beta 1 class VI; tubulin beta chain; tubulin beta class I; tubulin beta-1 chain; Tubulin beta-5 chain; tubulin, alpha 1; tubulin, alpha 1A; tubulin, alpha, brain-specific; tubulin, beta 1 class VI; tubulin, beta 2A class IIa; tubulin, beta class I; tubulin, beta family 1; tubulin, beta polypeptide; a/β-tubulin | |
Rabbit | |
Affinity chromatography | |
RUO | |
100134983, 100135051, 203068, 22142, 545486, 64158, 679312, 733686, 7846, 81027 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
A2AQ07, P02550, P07437, P68369, P68370, Q71U36, Q767L7, Q9H4B7 | |
TUBA1A, TUBB, TUBB1 | |
Purified porcine brain tubulin. | |
500 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction