Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUFM Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527512

Catalog No. PIPA527512


Add to cart

Description

Description

PA5-27512 targets TUFM in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27512 immunogen is recombit fragment corresponding to a region within amino acids 189 and 377 of Human TUFM. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a protein which participates in protein translation in mitochondria. Mutations in this gene have been associated with combined oxidative phosphorylation deficiency resulting in lactic acidosis and fatal encephalopathy. A pseudogene has been identified on chromosome 17.
Specifications

Specifications

TUFM
Polyclonal
Unconjugated
TUFM
Liquid
TUFM
Recombit fragment corresponding to a region within amino acids 189 and 377 of Human TUFM
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
P49411
COXPD4, EF-TuMT, EFTU, P43, EF-Tu, elongation factor Tu, mitochondrial
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7284
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit