Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-TURKEY IGG (H+L) AB FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300327
Affinity purified polyclonal antibody to Turkey IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-669-13