Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXK tyrosine kinase, Mouse, Clone: 2G10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TXK.
Supplier: Abnova Corporation H00007294M02
Description
Sequence: LIPSNYVTENKITNLEIYEWYHRNITRNQAEHLLRQESKEGAFIVRDSRHLGSYTISVFMGARRSTEAAIKHYQIKKNDSGQWYVAERHAFQSIPELIWYSpecifications
TXK tyrosine kinase | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003328 | |
TXK | |
TXK (NP_003319, 131 a.a. ∼ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Kinases | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
2G10 | |
Mouse monoclonal antibody raised against a partial recombinant TXK. | |
TXK | |
BTKL/MGC22473/PSCTK5/PTK4/RLK/TKL | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
7294 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction