Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524494

Catalog No. PIPA524494

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be a regulator of microtubule physiology.
Specifications

Specifications

TXNDC6
Polyclonal
PBS with 0.09% sodium azide
Q86XW9
TXL-2, TXL2, TXNDC6, thioredoxin domain containing 6, thioredoxin domain-containing protein 6, thioredoxin-like 2, thioredoxin-like protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
347736
Immunohistochemistry, Western Blot
Unconjugated
NME9
Liquid
NME9
KLH conjugated synthetic peptide between 111-139 amino acids from the central region of human TXNDC6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit