Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXNIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
$204.00 - $482.50
Specifications
Antigen | TXNIP |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2-4ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227095SS
|
Novus Biologicals
NBP227095SS |
0.025 mg |
Each for $204.00
|
|
|||||
NBP227095
|
Novus Biologicals
NBP227095 |
0.1 mg |
Each for $482.50
|
|
|||||
Description
TXNIP Polyclonal specifically detects TXNIP in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot.Specifications
TXNIP | |
Western Blot 2-4ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.05% Sodium Azide | |
HHCPA78, THIF, thioredoxin binding protein 2, thioredoxin interacting protein, Thioredoxin-binding protein 2, thioredoxin-interacting protein, upregulated by 1,25-dihydroxyvitamin D-3, VDUP1EST01027, Vitamin D3 up-regulated protein 1 | |
TXNIP | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 94% homologous in mouse and rat. |
1.0 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat, Primate, Monkey | |
Q9H3M7 | |
10628 | |
A portion of amino acids 100-150 of human TxNIP was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title