Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNL2, Polyclonal, Invitrogen™

Catalog No. PIPA127788
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127788 50 μg
1 options

Catalog No. PIPA127788

Supplier: Thermo Scientific PA127788

Chicken Polyclonal Antibody

May play a role in regulating the function of the thioredoxin system.

PA1-27788 detects TXNL2 from human, mouse, and rat samples.PA1-27788 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Thioredoxin-like protein 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNL2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene GLRX3
Gene Accession No. O76003, Q9CQM9, Q9JLZ1
Gene Alias FLJ11864, GLRX4, GRX3, GRX4, PICOT, TXNL2, TXNL3, bA500G10.4
Gene Symbols GLRX3
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Thioredoxin-like protein 2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10539
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.