Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRO3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522999
Description
The target sequence has 94% sequence homology with rat. Suggested positive control: normal human brain lysate, normal mouse brain lysate.
TYRO3 Polyclonal antibody specifically detects TYRO3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
TYRO3 | |
Polyclonal | |
Unconjugated | |
TYRO3 | |
AI323366; Brt; Bruton agammaglobulinemia tyrosine kinase; BYK; Dtk; Etk-2; etk2/tyro3; FLJ16467; Rek; Rse; Sky; soluble tyro 3; soluble tyro3; styro 3; styro3; TIF; TK19-2; tyro 3; TYRO3; TYRO3 protein tyrosine kinase; TYRO3 protein tyrosine kinase 3; tyrosine-protein kinase byk; Tyrosine-protein kinase DTK; tyrosine-protein kinase receptor TYRO3; Tyrosine-protein kinase RSE; tyrosine-protein kinase SKY; Tyrosine-protein kinase TIF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22174, 7301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
1.19 mg/mL | |
PBS with 30% glycerol and 0.1% sodium azide | |
P55144, Q06418 | |
TYRO3 | |
Synthetic peptide made to an internal portion of the human protein (within residues 100-250). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction