Learn More
Tyrosinase Mouse anti-Human, Clone: T311, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05647
Catalog No. 05-647-MI
Add to cart
Description
Description
This Anti-Tyrosinase Antibody, clone T311 is validated for use in western blot, immunohistochemistry for the detection of Tyrosinase.
Specifications
Specifications
Tyrosinase
T311
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
IgG2a
200μg
Primary
Human
Immunohistochemistry, Western Blot
Unconjugated
NM_000372
TYR, LB24-AB, tyrosinase, OCAIA, SK29-AB, OCA1A
Purified recombinant Tyrosinase corresponding to residues 5-456 of human Tyrosinase
Protein G chromatography
−20°C for 24 months.
Monoclonal